5-Brom-2-thiophencarbaldehyd

Lieferant: COMBI-BLOCKS

CA-4274-5G CA-4274-25G CA-4274-100G
COBBCA-4274-5GEA 0 EUR
COBBCA-4274-5G COBBCA-4274-25G COBBCA-4274-100G
5-Brom-2-thiophencarbaldehyd
5-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
MDL: MFCD00005432
CAS-Nummer: 4701-17-1
EINECS: 225-176-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR