2-Brom-5-methoxybenzoesäure

Lieferant: COMBI-BLOCKS

CA-4278-5G CA-4278-25G CA-4278-100G
COBBCA-4278-5GEA 0 EUR
COBBCA-4278-5G COBBCA-4278-25G COBBCA-4278-100G
2-Brom-5-methoxybenzoesäure
2-Bromo-5-methoxybenzoic acid
Formel: C₈H₇BrO₃
MDL: MFCD00020214
CAS-Nummer: 22921-68-2
EINECS: 245-329-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR