Methyl-4-brom-3-methylbenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-bromo-3-methylbenzoate
CA-4280-100G
CA-4280-5G
CA-4280-25G
COBBCA-4280-100GEA
312
EUR
COBBCA-4280-100G
COBBCA-4280-5G
COBBCA-4280-25G
Methyl-4-brom-3-methylbenzoat
Methyl-4-brom-3-methylbenzoat
Formel:
C₉H₉BrO₂ Molecular Weight: 229.07 g/mol |
MDL:
MFCD00673014 CAS-Nummer: 148547-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...