Methyl-4-brom-3-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-bromo-3-methylbenzoate

CA-4280-100G CA-4280-5G CA-4280-25G
COBBCA-4280-100GEA 312 EUR
COBBCA-4280-100G COBBCA-4280-5G COBBCA-4280-25G
Methyl-4-brom-3-methylbenzoat
Methyl-4-brom-3-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229.07 g/mol
MDL: MFCD00673014
CAS-Nummer: 148547-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR