3-Hydroxy-2-nitropyridin
Lieferant: COMBI-BLOCKS
CA-4300-100G
COBBCA-4300-100GEA
0
EUR
COBBCA-4300-100G
3-Hydroxy-2-nitropyridin
3-Hydroxy-2-nitropyridine
Formel:
C₅H₄N₂O₃ Schmelzpunkt: 70 °C |
MDL:
MFCD00006258 CAS-Nummer: 15128-82-2 EINECS: 239-191-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...