3-Brom-5-(trifluormethyl)benzoesäure

Lieferant: COMBI-BLOCKS

CA-4351-25G CA-4351-100G CA-4351-5G
COBBCA-4351-25GEA 0 EUR
COBBCA-4351-25G COBBCA-4351-100G COBBCA-4351-5G
3-Brom-5-(trifluormethyl)benzoesäure
3-Bromo-5-(trifluoromethyl)benzoic acid
Formel: C₈H₄BrF₃O₂
MDL: MFCD03412186
CAS-Nummer: 328-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR