N-(3-Brom-5-nitrobenzoyl)morpholin

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-5-nitrophenyl)(morpholino)methanone

CA-4377-1G CA-4377-5G
COBBCA-4377-1GEA 0 EUR
COBBCA-4377-1G COBBCA-4377-5G
N-(3-Brom-5-nitrobenzoyl)morpholin
N-(3-Brom-5-nitrobenzoyl)morpholin
Formel: C₁₁H₁₁BrN₂O₄
MDL: MFCD09475915
CAS-Nummer: 941294-19-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR