3-Brom-N-methylbenzamid

Lieferant: COMBI-BLOCKS

CA-4392-5G CA-4392-1G
COBBCA-4392-5GEA 514 EUR
COBBCA-4392-5G COBBCA-4392-1G
3-Brom-N-methylbenzamid
3-Brom-N-methylbenzamid
Formel: C₈H₈BrNO
Molecular Weight: 214.0592 g/mol
MDL: MFCD01060879
CAS-Nummer: 49834-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR