2-Brom-5-(trifluormethyl)benzonitril

Lieferant: COMBI-BLOCKS

CA-4407-25G CA-4407-5G CA-4407-100G
COBBCA-4407-25GEA 0 EUR
COBBCA-4407-25G COBBCA-4407-5G COBBCA-4407-100G
2-Brom-5-(trifluormethyl)benzonitril
2-Brom-5-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
MDL: MFCD03412187
CAS-Nummer: 1483-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR