2-Brom-5-(trifluormethyl)benzonitril
Lieferant: COMBI-BLOCKS
CA-4407-25G
CA-4407-5G
CA-4407-100G
COBBCA-4407-25GEA
0
EUR
COBBCA-4407-25G
COBBCA-4407-5G
COBBCA-4407-100G
2-Brom-5-(trifluormethyl)benzonitril
2-Brom-5-(trifluormethyl)benzonitril
Formel:
C₈H₃BrF₃N |
MDL:
MFCD03412187 CAS-Nummer: 1483-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...