2-Brom-N,N-dimethyl-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-5-fluoro-N,N-dimethylbenzamide

CA-4431-5G CA-4431-1G
COBBCA-4431-5GEA 1130 EUR
COBBCA-4431-5G COBBCA-4431-1G
2-Brom-N,N-dimethyl-5-fluorbenzamid
2-Brom-N,N-dimethyl-5-fluorbenzamid
Formel: C₉H₉BrFNO
Molecular Weight: 246.0763 g/mol
MDL: MFCD09800904
CAS-Nummer: 951884-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR