2-Brom-N-cyclohexyl-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4446-5G CA-4446-1G
COBBCA-4446-5GEA 0 EUR
COBBCA-4446-5G COBBCA-4446-1G
2-Brom-N-cyclohexyl-5-fluorbenzamid
2-Brom-N-cyclohexyl-5-fluorbenzamid
Formel: C₁₃H₁₅BrFNO
MDL: MFCD09800915
CAS-Nummer: 951884-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR