Methyl-3-brom-4-chlorbenzoat
Lieferant: COMBI-BLOCKS
CA-4472-5G
CA-4472-1G
COBBCA-4472-5GEA
0
EUR
COBBCA-4472-5G
COBBCA-4472-1G
Methyl-3-brom-4-chlorbenzoat
Methyl-3-brom-4-chlorbenzoat
Formel:
C₈H₆BrClO₂ |
MDL:
MFCD09751943 CAS-Nummer: 107947-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...