(±)-Indolin-2-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Indoline-2-carboxylic acid
CA-4489-5G
CA-4489-25G
CA-4489-100G
COBBCA-4489-5GEA
0
EUR
COBBCA-4489-5G
COBBCA-4489-25G
COBBCA-4489-100G
(±)-Indolin-2-carbonsäure
(±)-Indolin-2-carbonsäure
Formel:
C₉H₉NO₂ |
MDL:
MFCD00010635 CAS-Nummer: 78348-24-0 EINECS: 278-899-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...