Methyl-4-fluor-3-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-fluoro-3-methylbenzoate

CA-4506-1G
COBBCA-4506-1GEA 38.2 EUR
COBBCA-4506-1G
Methyl-4-fluor-3-methylbenzoat
Methyl-4-fluor-3-methylbenzoat
Formel: C₉H₉FO₂
Molecular Weight: 168,1650 g/mol
MDL: MFCD06203783
CAS-Nummer: 180636-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR