2-Fluor-m-toluonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-3-methylbenzonitrile

CA-4507-1G
COBBCA-4507-1GEA 25.5 EUR
COBBCA-4507-1G
2-Fluor-m-toluonitril
2-Fluor-m-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,1384 g/mol
MDL: MFCD07782078
CAS-Nummer: 185147-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR