2-Fluor-3-hydroxybenzonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluor-3-hydroxybenzoesäurenitril

CA-4540-1G
COBBCA-4540-1GEA 336 EUR
COBBCA-4540-1G
2-Fluor-3-hydroxybenzonitril
2-Fluor-3-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137,11 g/mol
MDL: MFCD09839222
CAS-Nummer: 1000339-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR