1-(4-Chlorbenzoyl)piperidin

Lieferant: COMBI-BLOCKS

CA-4545-5G CA-4545-25G CA-4545-1G
COBBCA-4545-5GEA 0 EUR
COBBCA-4545-5G COBBCA-4545-25G COBBCA-4545-1G
1-(4-Chlorbenzoyl)piperidin
1-(4-Chlorbenzoyl)piperidin
Formel: C₁₂H₁₄ClNO
MDL: MFCD00584705
CAS-Nummer: 26163-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR