4-Brom-N-cyclohexyl-3-methylbenzamid

Lieferant: COMBI-BLOCKS

CA-4563-5G CA-4563-1G
COBBCA-4563-5GEA 0 EUR
COBBCA-4563-5G COBBCA-4563-1G
4-Brom-N-cyclohexyl-3-methylbenzamid
4-Brom-N-cyclohexyl-3-methylbenzamid
Formel: C₁₄H₁₈BrNO
MDL: MFCD09972128
CAS-Nummer: 1020252-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR