Methyl-3-brom-5-(trifluormethyl)benzoat

Lieferant: COMBI-BLOCKS

CA-4592-5G CA-4592-25G CA-4592-1G
COBBCA-4592-5GEA 0 EUR
COBBCA-4592-5G COBBCA-4592-25G COBBCA-4592-1G
Methyl-3-brom-5-(trifluormethyl)benzoat
Methyl-3-brom-5-(trifluormethyl)benzoat
Formel: C₉H₆BrF₃O₂
MDL: MFCD11847338
CAS-Nummer: 187331-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR