N-(Furan-2-ylmethyl)-3-brom-4-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4633-5G CA-4633-25G CA-4633-1G
COBBCA-4633-5GEA 141 EUR
COBBCA-4633-5G COBBCA-4633-25G COBBCA-4633-1G
N-(Furan-2-ylmethyl)-3-brom-4-fluorbenzamid
N-(Furan-2-ylmethyl)-3-brom-4-fluorbenzamid
Formel: C₁₂H₉BrFNO₂
MDL: MFCD11053851
CAS-Nummer: 1065074-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR