4-Brom-N-cyclohexyl-3-methoxybenzamid

Lieferant: COMBI-BLOCKS

CA-4651-1G CA-4651-5G
COBBCA-4651-1GEA 0 EUR
COBBCA-4651-1G COBBCA-4651-5G
4-Brom-N-cyclohexyl-3-methoxybenzamid
4-Brom-N-cyclohexyl-3-methoxybenzamid
Formel: C₁₄H₁₈BrNO₂
MDL: MFCD11504875
CAS-Nummer: 1072944-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR