3-(3-Bromphenyl)propionsäure

Lieferant: COMBI-BLOCKS

CA-4673-25G CA-4673-1G CA-4673-5G
COBBCA-4673-25GEA 246 EUR
COBBCA-4673-25G COBBCA-4673-1G COBBCA-4673-5G
3-(3-Bromphenyl)propionsäure
3-(3-Bromphenyl)propionsäure
Formel: C₉H₉BrO₂
Molecular Weight: 229.07 g/mol
MDL: MFCD01310792
CAS-Nummer: 42287-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR