2-Brom-4-methoxybenzonitril

Lieferant: COMBI-BLOCKS

CA-4680-1G
COBBCA-4680-1GEA 57 EUR
COBBCA-4680-1G
2-Brom-4-methoxybenzonitril
2-Brom-4-methoxybenzonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,0433 g/mol
MDL: MFCD09056764
CAS-Nummer: 140860-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR