Methyl-4-methyl-3-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-methyl-3-nitrobenzoate

CA-4707-5G CA-4707-25G CA-4707-100G
COBBCA-4707-5GEA 0 EUR
COBBCA-4707-5G COBBCA-4707-25G COBBCA-4707-100G
Methyl-4-methyl-3-nitrobenzoat
Methyl-4-methyl-3-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195.18 g/mol
MDL: MFCD00130054
CAS-Nummer: 7356-11-8
EINECS: 230-886-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR