5-Brom-2-chlor-N-cyclopropylbenzamid

Lieferant: COMBI-BLOCKS

CA-4723-5G CA-4723-1G
COBBCA-4723-5GEA 0 EUR
COBBCA-4723-5G COBBCA-4723-1G
5-Brom-2-chlor-N-cyclopropylbenzamid
5-Brom-2-chlor-N-cyclopropylbenzamid
Formel: C₁₀H₉BrClNO
MDL: MFCD04226747
CAS-Nummer: 669734-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR