1-(4-Brom-2-chlorbenzoyl)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromo-2-chlorophenyl)(piperidin-1-yl)methanone

CA-4738-1G CA-4738-5G
COBBCA-4738-1GEA 149 EUR
COBBCA-4738-1G COBBCA-4738-5G
1-(4-Brom-2-chlorbenzoyl)piperidin
1-(4-Brom-2-chlorbenzoyl)piperidin
Formel: C₁₂H₁₃BrClNO
MDL: MFCD12756428
CAS-Nummer: 1187385-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR