N-Cyclohexyl-3-fluorbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N-Cyclohexyl-3-fluorobenzamide

CA-4768-5G
COBBCA-4768-5GEA 149 EUR
COBBCA-4768-5G
N-Cyclohexyl-3-fluorbenzamid
N-Cyclohexyl-3-fluorbenzamid
Formel: C₁₃H₁₆FNO
Molecular Weight: 221.270 g/mol
MDL: MFCD00454568
CAS-Nummer: 2267-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR