Methyl-2-brom-6-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromo-6-methylbenzoate

CA-4787-1G
COBBCA-4787-1GEA 38.2 EUR
COBBCA-4787-1G
Methyl-2-brom-6-methylbenzoat
Methyl-2-brom-6-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,0700 g/mol
MDL: MFCD09260938
CAS-Nummer: 99548-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR