Methyl-4-brombutyrat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-bromobutanoate

CA-4808-25G CA-4808-100G CA-4808-5G
COBBCA-4808-25GEA 38.2 EUR
COBBCA-4808-25G COBBCA-4808-100G COBBCA-4808-5G
Methyl-4-brombutyrat
Methyl-4-brombutyrat
Formel: C₅H₉BrO₂
Molecular Weight: 181.03 g/mol
MDL: MFCD00041482
CAS-Nummer: 4897-84-1
EINECS: 225-523-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR