3'-Hydroxybiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3'-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid
CA-4816-1G
CA-4816-5G
COBBCA-4816-1GEA
452
EUR
COBBCA-4816-1G
COBBCA-4816-5G
3'-Hydroxybiphenyl-3-carbonsäure
3'-Hydroxybiphenyl-3-carbonsäure
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214.22 g/mol |
MDL:
MFCD04117370 CAS-Nummer: 171047-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...