4'-Hydroxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4817-1G
COBBCA-4817-1GEA 385 EUR
COBBCA-4817-1G
4'-Hydroxybiphenyl-3-carbonsäure
4'-Hydroxybiphenyl-3-carbonsäure
Formel: C₁₃H₁₀O₃
MDL: MFCD01910126
CAS-Nummer: 121629-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR