2'-Methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4819-5G CA-4819-1G
COBBCA-4819-5GEA 525 EUR
COBBCA-4819-5G COBBCA-4819-1G
2'-Methoxybiphenyl-3-carbonsäure
2'-Methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD03424597
CAS-Nummer: 168618-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR