2'-Nitrobiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4833-1G
COBBCA-4833-1GEA 303 EUR
COBBCA-4833-1G
2'-Nitrobiphenyl-3-carbonsäure
2'-Nitrobiphenyl-3-carbonsäure
Formel: C₁₃H₉NO₄
MDL: MFCD00834942
CAS-Nummer: 136102-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR