2'-Aminobiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4867-1G
COBBCA-4867-1GEA 437 EUR
COBBCA-4867-1G
2'-Aminobiphenyl-3-carbonsäure
2'-Aminobiphenyl-3-carbonsäure
Formel: C₁₃H₁₁NO₂
MDL: MFCD03990478
CAS-Nummer: 177171-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR