2'-Acetylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4869-1G
COBBCA-4869-1GEA 404 EUR
COBBCA-4869-1G
2'-Acetylbiphenyl-3-carbonsäure
2'-Acetylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₂O₃
MDL: MFCD06802324
CAS-Nummer: 870245-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR