2'-tert-Butylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4876-1G
COBBCA-4876-1GEA 1110 EUR
COBBCA-4876-1G
2'-tert-Butylbiphenyl-3-carbonsäure
2'-tert-Butylbiphenyl-3-carbonsäure
Formel: C₁₇H₁₈O₂
MDL: MFCD00795837
CAS-Nummer: 40182-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR