2'-Sulphamoylbiphenyl-3-carboxylic acid

Lieferant: COMBI-BLOCKS

CA-4879-1G
COBBCA-4879-1GEA 0 EUR
COBBCA-4879-1G
2'-Sulphamoylbiphenyl-3-carboxylic acid
2'-Sulphamoylbiphenyl-3-carboxylic acid
Formel: C₁₃H₁₁NO₄S
MDL: MFCD15143493
CAS-Nummer: 1215206-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR