4'-(Trifluormethoxy)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4892-1G
COBBCA-4892-1GEA 281 EUR
COBBCA-4892-1G
4'-(Trifluormethoxy)biphenyl-3-carbonsäure
4'-(Trifluormethoxy)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₃
Molecular Weight: 282,22 g/mol
MDL: MFCD04117416
CAS-Nummer: 728919-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR