2',3'-Dimethylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4897-1G
COBBCA-4897-1GEA 864 EUR
COBBCA-4897-1G
2',3'-Dimethylbiphenyl-3-carbonsäure
2',3'-Dimethylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₂
MDL: MFCD15143499
CAS-Nummer: 1215206-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR