2'-Chlor-4'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4900-1G
COBBCA-4900-1GEA 864 EUR
COBBCA-4900-1G
2'-Chlor-4'-methylbiphenyl-3-carbonsäure
2'-Chlor-4'-methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₂
MDL: MFCD15143500
CAS-Nummer: 1215206-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR