2'-Chlor-6'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4901-1G
COBBCA-4901-1GEA 1110 EUR
COBBCA-4901-1G
2'-Chlor-6'-methylbiphenyl-3-carbonsäure
2'-Chlor-6'-methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₂
MDL: MFCD15143501
CAS-Nummer: 1215206-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR