2'-Brom-3'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4904-1G
COBBCA-4904-1GEA 864 EUR
COBBCA-4904-1G
2'-Brom-3'-methylbiphenyl-3-carbonsäure
2'-Brom-3'-methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291.14 g/mol
MDL: MFCD15143504
CAS-Nummer: 1215206-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR