2'-Brom-5'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4906-1G
COBBCA-4906-1GEA 864 EUR
COBBCA-4906-1G
2'-Brom-5'-methylbiphenyl-3-carbonsäure
2'-Brom-5'-methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₁BrO₂
MDL: MFCD15143506
CAS-Nummer: 1215206-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR