2'-Methoxy-3'-methylbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4913-1G
COBBCA-4913-1GEA
864
EUR
COBBCA-4913-1G
2'-Methoxy-3'-methylbiphenyl-3-carbonsäure
2'-Methoxy-3'-methylbiphenyl-3-carbonsäure
Formel:
C₁₅H₁₄O₃ |
MDL:
MFCD15143512 CAS-Nummer: 1215206-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...