2'-Methoxy-4'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4915-1G
COBBCA-4915-1GEA 864 EUR
COBBCA-4915-1G
2'-Methoxy-4'-methylbiphenyl-3-carbonsäure
2'-Methoxy-4'-methylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₃
MDL: MFCD15143513
CAS-Nummer: 1215205-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR