2',3'-Dichlorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4916-1G
COBBCA-4916-1GEA 864 EUR
COBBCA-4916-1G
2',3'-Dichlorbiphenyl-3-carbonsäure
2',3'-Dichlorbiphenyl-3-carbonsäure
Formel: C₁₃H₈Cl₂O₂
MDL: MFCD11932367
CAS-Nummer: 177734-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR