2',6'-Dichlorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4918-1G
COBBCA-4918-1GEA 0 EUR
COBBCA-4918-1G
2',6'-Dichlorbiphenyl-3-carbonsäure
2',6'-Dichlorbiphenyl-3-carbonsäure
Formel: C₁₃H₈Cl₂O₂
MDL: MFCD11932350
CAS-Nummer: 1181345-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR