3'-Methoxy-2'-methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4920-1G
COBBCA-4920-1GEA 864 EUR
COBBCA-4920-1G
3'-Methoxy-2'-methylbiphenyl-3-carbonsäure
3'-Methoxy-2'-methylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₃
MDL: MFCD15143514
CAS-Nummer: 1215206-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR