2'-Chlor-3'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4924-1G
COBBCA-4924-1GEA 864 EUR
COBBCA-4924-1G
2'-Chlor-3'-methoxybiphenyl-3-carbonsäure
2'-Chlor-3'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁ClO₃
MDL: MFCD15143518
CAS-Nummer: 1215206-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR